ContactUsLinkComponent
QuenchShield Mountant is glycerol based aqueous mountant, which prevents quenching of fluorescence of fluorescent labeled cells and tissues samples during fluorescence microscopy experiments. In addition, it helps preserve the fluorescence of store 1 * 2 mL
Catalog # GENO786-1269
Supplier:  G-Biosciences
QuenchShield Mountant is glycerol based aqueous mountant, which prevents quenching of fluorescence of fluorescent labeled cells and tissues samples during fluorescence microscopy experiments. In addition, it helps preserve the fluorescence of store 1 * 2 mL
Catalog # GENO786-1269
Supplier:  G-Biosciences
Frequently Bought Together