ContactUsLinkComponent
Trypsin is a serine protease that cleaves peptides on C-terminal end of lysine and arginine amino acid residues. The pH optimum of t rypsin is pH 7.0-8.0. 1 * 50 mg
Catalog # GENO786-1255
Supplier:  G-Biosciences
Trypsin is a serine protease that cleaves peptides on C-terminal end of lysine and arginine amino acid residues. The pH optimum of t rypsin is pH 7.0-8.0. 1 * 50 mg
Catalog # GENO786-1255
Supplier:  G-Biosciences
Frequently Bought Together