ContactUsLinkComponent
Trypsin is a serine protease that cleaves peptides on C-terminal end of lysine and arginine amino acid residues. The pH optimum of t rypsin is pH 7.0-8.0. 1 * 5 mg
  GENO786-1254
Trypsin is a serine protease that cleaves peptides on C-terminal end of lysine and arginine amino acid residues. The pH optimum of t rypsin is pH 7.0-8.0. 1 * 5 mg
  GENO786-1254
Frequently Bought Together