Trypsin is a serine protease that cleaves peptides on C-terminal end of lysine and arginine amino acid residues. The pH optimum of t rypsin is pH 7.0-8.0. 1 * 1 mg
Catalog # GENO786-1253
Supplier: G-Biosciences
Trypsin is a serine protease that cleaves peptides on C-terminal end of lysine and arginine amino acid residues. The pH optimum of t rypsin is pH 7.0-8.0. 1 * 1 mg
Catalog # GENO786-1253
Supplier: G-Biosciences
Frequently Bought Together
Chloroform ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Dichloromethane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
2-Propanol, dehydrated (max. 0.01% H₂O) ≥99.8%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabiliser free, AcroSeal®
Supplier: Thermo Fisher Scientific
Petroleum spirit, 60…80 °C
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.8%, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor