ContactUsLinkComponent
SG-Carboxypeptidase B (Recombinant)is the rat carboxypeptidase B expressed in E. coli. Carboxypeptidase B specifically hydrolyses b asic amino acids including lysine, arginine and histidine from the C-terminal end of polypeptides. It is used in sequ 1 * 1 mg
Catalog # GENO786-1250
Supplier:  G-Biosciences
SG-Carboxypeptidase B (Recombinant)is the rat carboxypeptidase B expressed in E. coli. Carboxypeptidase B specifically hydrolyses b asic amino acids including lysine, arginine and histidine from the C-terminal end of polypeptides. It is used in sequ 1 * 1 mg
Catalog # GENO786-1250
Supplier:  G-Biosciences
Frequently Bought Together