ContactUsLinkComponent
Sputo-LR™ is a sputum liquefying reagent for sputum and other mucoid fluids that is useful for the isolation of pathogenic or saprop hytic organisms or cells. A concentrated sterile solution of Cleland’s Reagent (DTT) in phosphate buffer at pH 7.0. 1 * 10 mL
Catalog # GENO786-1086
Supplier:  G-Biosciences
Sputo-LR™ is a sputum liquefying reagent for sputum and other mucoid fluids that is useful for the isolation of pathogenic or saprop hytic organisms or cells. A concentrated sterile solution of Cleland’s Reagent (DTT) in phosphate buffer at pH 7.0. 1 * 10 mL
Catalog # GENO786-1086
Supplier:  G-Biosciences
Frequently Bought Together