Aqueous Eosin Y Solution [1%], a conventional stain used for Hematoxylin and Eosin (H&E) staining in histology and cytology procedur es. Ready to use formulation optimized for histology and cytology procedures where reproducible results are important 1 * 500 mL
Catalog # GENO786-1072
Supplier: G-Biosciences
Frequently Bought Together