ContactUsLinkComponent
Apoptosis is a form of programmed cell death which is essential as a normal part of development in multicellular organisms. Apoptosi s go awry in some disease states such as Alzheimer’s and cancer. Annexin V-FITC Apoptosis Assay is designed to detect 1 * 20 Assays
Catalog # GENO786-1548
Supplier:  G-Biosciences
Apoptosis is a form of programmed cell death which is essential as a normal part of development in multicellular organisms. Apoptosi s go awry in some disease states such as Alzheimer’s and cancer. Annexin V-FITC Apoptosis Assay is designed to detect 1 * 20 Assays
Catalog # GENO786-1548
Supplier:  G-Biosciences
Frequently Bought Together