ContactUsLinkComponent
G-Sep is a gel filtration resin comprised of ultrapure cross-linked dextran for desalting and buffer exchange in industrial applic ations. It exhibits high selectivity, superb resolution, low non-specific adsorption and robust chemical stability. 1 * 100 g
Catalog # GENO786-1393
Supplier:  G-Biosciences
G-Sep is a gel filtration resin comprised of ultrapure cross-linked dextran for desalting and buffer exchange in industrial applic ations. It exhibits high selectivity, superb resolution, low non-specific adsorption and robust chemical stability. 1 * 100 g
Catalog # GENO786-1393
Supplier:  G-Biosciences
Frequently Bought Together