ContactUsLinkComponent
G-Sep is a gel filtration resin comprised of ultrapure cross-linked dextran for desalting and buffer exchange in industrial applica tions. It exhibits high selectivity, superb resolution, low non-specific adsorption and robust chemical stability. 1 * 1 kg
  GENO786-1392
G-Sep is a gel filtration resin comprised of ultrapure cross-linked dextran for desalting and buffer exchange in industrial applica tions. It exhibits high selectivity, superb resolution, low non-specific adsorption and robust chemical stability. 1 * 1 kg
  GENO786-1392
Frequently Bought Together