ContactUsLinkComponent
Lumino Firefly Luciferase Glow Assay is designed for high throughput screening or quantification of luciferase expression in mammal ian cells which is mainly performed by batch processing of 96- and 384- well plates. 1 * 100 Assays
Catalog # GENO786-1353
Supplier:  G-Biosciences
Lumino Firefly Luciferase Glow Assay is designed for high throughput screening or quantification of luciferase expression in mammal ian cells which is mainly performed by batch processing of 96- and 384- well plates. 1 * 100 Assays
Catalog # GENO786-1353
Supplier:  G-Biosciences
Frequently Bought Together