ContactUsLinkComponent
JC-1 Mitochondrial Membrane Potential Assay comprise of lipophilic cationic fluorescent dye. In healthy cells, JC-1 enters the energ ized mitochondria and form aggregates which change the fluorescent property of JC-1 dye. Unhealthy or apoptotic cells 1 * 100 Tests
Catalog # GENO786-1322
Supplier:  G-Biosciences
JC-1 Mitochondrial Membrane Potential Assay comprise of lipophilic cationic fluorescent dye. In healthy cells, JC-1 enters the energ ized mitochondria and form aggregates which change the fluorescent property of JC-1 dye. Unhealthy or apoptotic cells 1 * 100 Tests
Catalog # GENO786-1322
Supplier:  G-Biosciences
Frequently Bought Together