ContactUsLinkComponent
FCCP [Carbonyl cyanide 4-(trifluoromethoxy) phenylhydrazone] is a potent uncoupler of oxidative phosphorylation in mitochondria. It is used as a positive control for mitochondrial membrane depolarization. FCCP disrupts the synthesis of ATP by trnas 1 * 50 mg
  GENO786-1317
FCCP [Carbonyl cyanide 4-(trifluoromethoxy) phenylhydrazone] is a potent uncoupler of oxidative phosphorylation in mitochondria. It is used as a positive control for mitochondrial membrane depolarization. FCCP disrupts the synthesis of ATP by trnas 1 * 50 mg
  GENO786-1317
Frequently Bought Together