ContactUsLinkComponent
Lumino™ ATP Detection Assay is designed for rapid, convenient and quantitative detection of adenosine-5’-triphosphate (ATP). It’s a bioluminescent assay which is based on requirement of ATP by luciferase enzyme for production of light. 1 * 50 Assays
Catalog # GENO786-1311
Supplier:  G-Biosciences
Lumino™ ATP Detection Assay is designed for rapid, convenient and quantitative detection of adenosine-5’-triphosphate (ATP). It’s a bioluminescent assay which is based on requirement of ATP by luciferase enzyme for production of light. 1 * 50 Assays
Catalog # GENO786-1311
Supplier:  G-Biosciences
Frequently Bought Together