ContactUsLinkComponent
Quick release / Coupling M24x1,5 Temperature range: -75°C...230°C Connection: M24X1,5 AG Connection point: Tempering hose max. pressure at 20°C: 25 bar Nominal size: 12 mm Note: When using ethanol as thermofluid: When ethanol is used as thermofluid: 1 * 1 items
Catalog # HUBE10530
Supplier:  HUBER
Quick release / Coupling M24x1,5 Temperature range: -75°C...230°C Connection: M24X1,5 AG Connection point: Tempering hose max. pressure at 20°C: 25 bar Nominal size: 12 mm Note: When using ethanol as thermofluid: When ethanol is used as thermofluid: 1 * 1 items
Catalog # HUBE10530
Supplier:  HUBER
Frequently Bought Together