ContactUsLinkComponent
Quick release / nipple M24x1,5 Temperature range: -75°C...230°C Connection: M24X1,5 AG Connection point: Temperature control unit, application max. pressure at 20°C: 25 bar Nominal width: 12 mm Note: When using ethanol as thermofluid: When ethanol is 1 * 1 items
Catalog # HUBE10529
Supplier:  HUBER
Quick release / nipple M24x1,5 Temperature range: -75°C...230°C Connection: M24X1,5 AG Connection point: Temperature control unit, application max. pressure at 20°C: 25 bar Nominal width: 12 mm Note: When using ethanol as thermofluid: When ethanol is 1 * 1 items
Catalog # HUBE10529
Supplier:  HUBER
Frequently Bought Together