ContactUsLinkComponent
Caenorhabditis elegans is a soil nematode with great potential for educational research, partly because of its rapid (3-day) life cy cle, small size (1.0-mm-long adult), and ease of laboratory growth cultivation. Maximum Shelf Life: 9 Months(Worms 2 1 * 1 KIT
Catalog # EDVO856
Supplier:  EDVOTEK
Caenorhabditis elegans is a soil nematode with great potential for educational research, partly because of its rapid (3-day) life cy cle, small size (1.0-mm-long adult), and ease of laboratory growth cultivation. Maximum Shelf Life: 9 Months(Worms 2 1 * 1 KIT
Catalog # EDVO856
Supplier:  EDVOTEK
Frequently Bought Together