ContactUsLinkComponent
Welch Piston Pump, 230V 50/60Hz Ultinate pressure 100 Torr Pumping speed 50 Hz 18 l/min Motor power 93 W IN/EX hose connector DN 8 1 * 1 items
Catalog # ILMVU2522-C02
Welch Piston Pump, 230V 50/60Hz Ultinate pressure 100 Torr Pumping speed 50 Hz 18 l/min Motor power 93 W IN/EX hose connector DN 8 1 * 1 items
Catalog # ILMVU2522-C02
Frequently Bought Together