ContactUsLinkComponent
GreenGel cup 1% TBE. Re-meltable agarose gel with fluorescent DNA stain in TBE buffer for 1% gels. Includes also 100ml TBE concentra te. 1 * 10 items
  EMBIM3102TBE
 :  EMBITEC
GreenGel cup 1% TBE. Re-meltable agarose gel with fluorescent DNA stain in TBE buffer for 1% gels. Includes also 100ml TBE concentra te. 1 * 10 items
  EMBIM3102TBE
 :  EMBITEC
Frequently Bought Together