ContactUsLinkComponent
MiniLab PCR 101 and gel electrophoresis. Includes lambda phage DNA template (275µl), FastTaq PCR MasterMix (475µl), three primer set s (80µl each), MiniOne DNA marker (130µl), sterile nuclease-free water (80µl), MiniOne 5X sample loading dye (275µl), 1 * 1 KIT
Catalog # EMBIM6001
Supplier:  EMBITEC
MiniLab PCR 101 and gel electrophoresis. Includes lambda phage DNA template (275µl), FastTaq PCR MasterMix (475µl), three primer set s (80µl each), MiniOne DNA marker (130µl), sterile nuclease-free water (80µl), MiniOne 5X sample loading dye (275µl), 1 * 1 KIT
Catalog # EMBIM6001
Supplier:  EMBITEC
Frequently Bought Together