ContactUsLinkComponent
MiniLab PCR 101 amplification from the lambda phage genome. Includes lambda phage DNA template (275µl), FastTaq PCR MasterMix (475µl ), three primer sets (80µl each), MiniOne DNA marker (130µl), sterile nuclease-free water (80µl), MiniOne 5X sample l 1 * 1 KIT
Catalog # EMBIM6002
Supplier:  EMBITEC
MiniLab PCR 101 amplification from the lambda phage genome. Includes lambda phage DNA template (275µl), FastTaq PCR MasterMix (475µl ), three primer sets (80µl each), MiniOne DNA marker (130µl), sterile nuclease-free water (80µl), MiniOne 5X sample l 1 * 1 KIT
Catalog # EMBIM6002
Supplier:  EMBITEC
Frequently Bought Together