ContactUsLinkComponent
MiniLab PTC Genetics. Includes DNA Samples (6), GreenGel cups (10), TBE concentrate (100ml), 0,65ml microcentrifuge tubes (66 tubes) , micropipette tips (70 tips) for ten groups of students. Teacher and student guides. 1 * 1 KIT
Catalog # EMBIM3003
Supplier:  EMBITEC
MiniLab PTC Genetics. Includes DNA Samples (6), GreenGel cups (10), TBE concentrate (100ml), 0,65ml microcentrifuge tubes (66 tubes) , micropipette tips (70 tips) for ten groups of students. Teacher and student guides. 1 * 1 KIT
Catalog # EMBIM3003
Supplier:  EMBITEC
Frequently Bought Together