ContactUsLinkComponent
Film/Filter Holder for GENESYS 100 series. Accommodates filters/lenses up to 50 mm long x 50 mm tall x 1-8 mm thick. Comes assem bled in a removable sample compartment liner. 1 * 1 items
Catalog # 634-1097
Film/Filter Holder for GENESYS 100 series. Accommodates filters/lenses up to 50 mm long x 50 mm tall x 1-8 mm thick. Comes assem bled in a removable sample compartment liner. 1 * 1 items
Catalog # 634-1097
Frequently Bought Together