ContactUsLinkComponent
Color screen potentiometric titrator supplied with one burette driver assembly, 25 mL glass burette, and propeller stirrer, 240V 1 * 1 items
Catalog # HANNHI901C-02
Supplier:  Hanna
Color screen potentiometric titrator supplied with one burette driver assembly, 25 mL glass burette, and propeller stirrer, 240V 1 * 1 items
Catalog # HANNHI901C-02
Supplier:  Hanna
Frequently Bought Together