ContactUsLinkComponent
Stericlin® barcode-labels, on roll, for thermal transfer printers, paper, double self-adhesive (sandwich), with STEAM indicator, rol l width 70mm x length 80m x core 50mm 1 * 5 Roll
Catalog # VEPA3FSKS675105
Stericlin® barcode-labels, on roll, for thermal transfer printers, paper, double self-adhesive (sandwich), with STEAM indicator, rol l width 70mm x length 80m x core 50mm 1 * 5 Roll
Catalog # VEPA3FSKS675105
Frequently Bought Together