ContactUsLinkComponent
Easy Purchase Pack, includes (100) 12x32mm, 300ul screw top autosampler vials that are made from translucent ultra virgin polypropyl ene and (100) blue screw caps with a tan silicone/PTFE pre-slit septa. AQ Brand. 1 * 100 items
Catalog # MICS9532S-MS-3PP
Supplier:  MICROSOLV
Easy Purchase Pack, includes (100) 12x32mm, 300ul screw top autosampler vials that are made from translucent ultra virgin polypropyl ene and (100) blue screw caps with a tan silicone/PTFE pre-slit septa. AQ Brand. 1 * 100 items
Catalog # MICS9532S-MS-3PP
Supplier:  MICROSOLV
Frequently Bought Together