ContactUsLinkComponent
Easy Purchase Pack, with (100) 300ul clear fused insert (12x32mm) screw top vials with write on patch made with RSA-Pro glass and ( 100) light blue AQR pre-slit screw caps with fitted Silicone/PTFE septa. RSA-Pro Brand. 1 * 100 items
Catalog # MICS9532S-3DCP-MR
Supplier:  MICROSOLV
Easy Purchase Pack, with (100) 300ul clear fused insert (12x32mm) screw top vials with write on patch made with RSA-Pro glass and ( 100) light blue AQR pre-slit screw caps with fitted Silicone/PTFE septa. RSA-Pro Brand. 1 * 100 items
Catalog # MICS9532S-3DCP-MR
Supplier:  MICROSOLV
Frequently Bought Together