ContactUsLinkComponent
Assembly collar piston, 5 mm, piston for G7161A 1260 Infinity II preprative binary pump and G7161B 1290 Infinity II preprative binar y pump with AtP pump head 1 * 1 items
  HEWLG7161-60005
 :  Agilent
undefined
Assembly collar piston, 5 mm, piston for G7161A 1260 Infinity II preprative binary pump and G7161B 1290 Infinity II preprative binar y pump with AtP pump head 1 * 1 items
  HEWLG7161-60005
 :  Agilent
Frequently Bought Together