ContactUsLinkComponent
Agilent J+W DB-HeavyWAX GC column, 50 m, 0.32 mm, 0.20 µm 1 * 1 items
Catalog # HEWL123-7157
Supplier:  Agilent
Agilent J+W DB-HeavyWAX GC column, 50 m, 0.32 mm, 0.20 µm 1 * 1 items
Catalog # HEWL123-7157
Supplier:  Agilent
Frequently Bought Together