ContactUsLinkComponent
Agilent J+W DB-HeavyWAX GC column, 30 m, 0.25 mm, 0.50 µm 1 * 1 items
Catalog # HEWL122-7133
Supplier:  Agilent
undefined
Agilent J+W DB-HeavyWAX GC column, 30 m, 0.25 mm, 0.50 µm 1 * 1 items
Catalog # HEWL122-7133
Supplier:  Agilent
Frequently Bought Together