ContactUsLinkComponent
Skimmer cone, Nickel, for Agilent 7900/8900 Series ICP-MS with m-lens. Use with nickel sampler cone (G3280-67040) and stainless stee l skimmer base. Optional cone for use with m-lens. 1 * 1 items
Catalog # HEWLG3666-67511
Supplier:  Agilent
Skimmer cone, Nickel, for Agilent 7900/8900 Series ICP-MS with m-lens. Use with nickel sampler cone (G3280-67040) and stainless stee l skimmer base. Optional cone for use with m-lens. 1 * 1 items
Catalog # HEWLG3666-67511
Supplier:  Agilent
Frequently Bought Together