ContactUsLinkComponent
VACUETTE® VENOUS BLOOD COLLECTION TUBE PLASMA TUBE SODIUM HEPARIN ADDITIVE 13 X 100 MM 6 ML GREEN / BLACK RING PULL CAP POLYETHYLENE TEREPHTHALATE (PET) TUBE 1 * 1.200 items
Catalog # GREI456028_1200
Supplier:  Greiner Bio-One
VACUETTE® VENOUS BLOOD COLLECTION TUBE PLASMA TUBE SODIUM HEPARIN ADDITIVE 13 X 100 MM 6 ML GREEN / BLACK RING PULL CAP POLYETHYLENE TEREPHTHALATE (PET) TUBE 1 * 1.200 items
Catalog # GREI456028_1200
Supplier:  Greiner Bio-One
Frequently Bought Together