POTASSIUM HYDROXIDE 0,1 MOL/L (0,1N) IN 2-PROPANOL VOLUMETRIC SOLUTION 1 * 1 L
Katalogové číslo APPC183336.1611
Dodavatel: PanReac AppliChem
POTASSIUM HYDROXIDE 0,1 MOL/L (0,1N) IN 2-PROPANOL VOLUMETRIC SOLUTION 1 * 1 L
Katalogové číslo APPC183336.1611
Dodavatel: PanReac AppliChem
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Iodine 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Dodavatel: VWR Chemicals
VWR® Facial Tissues
Dodavatel: VWR Collection
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Dodavatel: Merck
pH indicator strips, MQuant®, Supelco®
Dodavatel: Merck
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Dodavatel: VWR Chemicals
VWR®, Beakers, Tall Form
Dodavatel: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals