ContactUsLinkComponent
Reaction tube 5ml polypropylene natural conical bottom graduated separate blue screw cap non-sterile (100 tubes/bag) 1 * 500 items
Catalog # GREI725201_500
Supplier:  Greiner Bio-One
Reaction tube 5ml polypropylene natural conical bottom graduated separate blue screw cap non-sterile (100 tubes/bag) 1 * 500 items
Catalog # GREI725201_500
Supplier:  Greiner Bio-One
Frequently Bought Together