ContactUsLinkComponent
Formaldehyde solution 4%, buffered, pH 6.9 (approx. 10% Formalin solution) for histology 1 * 700 mL
Catalog # 1.00496.0700
Supplier:  Merck
Formaldehyde solution 4%, buffered, pH 6.9 (approx. 10% Formalin solution) for histology 1 * 700 mL
Catalog # 1.00496.0700
Supplier:  Merck