ContactUsLinkComponent
Caps, blue, 9-425mm screw threads, bonded, preslit tan Silicone/PTFE septa. AQ brand, certified for LC & GC. 100/pack. 1 * 100 items
Catalog # MICS9502S-31-MB
Supplier:  MICROSOLV
undefined
Caps, blue, 9-425mm screw threads, bonded, preslit tan Silicone/PTFE septa. AQ brand, certified for LC & GC. 100/pack. 1 * 100 items
Catalog # MICS9502S-31-MB
Supplier:  MICROSOLV
Frequently Bought Together