Mouse Recombinant FGF-Basic (from E. coli)
Vial, 500 µG
Catalog # SHBT200-12-500UG
Supplier: Shenandoah Biotechnology
Mouse Recombinant FGF-Basic (from <i>E. coli</i>)
Catalog # SHBT200-12-500UG
Supplier: Shenandoah Biotechnology
CAS Number:
Jump to:
Product Details & Documents
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. The application of FGF-basic is a critical component for human embryonic stem cell culture systems and is necessary for maintaining human embryonic stem cells in an undifferentiated state.
- High quality
- Low endotoxin
- Affordable
: For research use only.
Specifications
Specifications
- Pack type:Vial
- Pk:500 µG
- Protease-free:N
- Purity:≥95%
- Source:E. coli
- Molecular weight:16,5 kDa
- Nuclease-free:N
- Protein/peptide type:Recombinant
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Protein function:Cytokine
- Gene ID:P15655
- Formulation:10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7,5
- Endotoxin-free:N
- Tested applications:Bioassay
- Species:Mouse
- Protein/peptide name:FGF-Basic
- Protein synonyms:Prostatropin|HBGF-2|FGF2
- Endotoxin level:Low
- Grade:RUO
- Sequence:MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- Biological activity:Bioactivity Assay: 3T3 cell proliferation, ED₅₀ ≤2,5 ng/ml, Bioactivity ≥4,0×10⁵ units/mg
- Shipping temperature:RT
- Cat. No.:SHBT200-12-500UG
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Storage conditions:–20 °C
- Carrier-free:Y
- Conjugation:Unconjugated
- Animal-free:N



