ContactUsLinkComponent
Paramount Ductless Enclosure, 3', 360 degree visibility with glass rear wall and baffle, 32 H x 29 D, 230V 50/60 Hz 1 * 1 items
Catalog # LBCP6963322
Supplier:  LABCONCO
Paramount Ductless Enclosure, 3', 360 degree visibility with glass rear wall and baffle, 32 H x 29 D, 230V 50/60 Hz 1 * 1 items
Catalog # LBCP6963322
Supplier:  LABCONCO
Frequently Bought Together