ContactUsLinkComponent
Male Nuts, HPLC, Combihead, Peek, 1 piece, Part knurled & part flat 12mm head. Finger tightening and or wrench. Total length 22mm, 8mm thread length, 10-32, blue color. 10/pack. 1 * 10 items
Catalog # MICS47320-11B
Supplier:  MICROSOLV
undefined
Male Nuts, HPLC, Combihead, Peek, 1 piece, Part knurled & part flat 12mm head. Finger tightening and or wrench. Total length 22mm, 8mm thread length, 10-32, blue color. 10/pack. 1 * 10 items
Catalog # MICS47320-11B
Supplier:  MICROSOLV
Frequently Bought Together