ContactUsLinkComponent
Ratiolab® Cryo-Boxes, PP, grid 9 x 9, blue, 133 x 133 x 50/75 mm, combi-lid, ,,numerically coded 1 * 5 items
Catalog # RATI5320014
Supplier:  RATIOLAB
Ratiolab® Cryo-Boxes, PP, grid 9 x 9, blue, 133 x 133 x 50/75 mm, combi-lid, ,,numerically coded 1 * 5 items
Catalog # RATI5320014
Supplier:  RATIOLAB
Frequently Bought Together