ContactUsLinkComponent
Antimicrobial illuminated Silicone keyboard Number of Keys: 110 Switch technology: Silicone keypad Protection level: IP68 Material, Colour: silicone, white Backlight: blue Interface: USB Switching cycles: 2 mill.(per key) Operating temperatur 1 * 1 items
Catalog # GETTKG24230
Supplier:  GETT GERATETECHNIK
Antimicrobial illuminated Silicone keyboard Number of Keys: 110 Switch technology: Silicone keypad Protection level: IP68 Material, Colour: silicone, white Backlight: blue Interface: USB Switching cycles: 2 mill.(per key) Operating temperatur 1 * 1 items
Catalog # GETTKG24230
Supplier:  GETT GERATETECHNIK
Frequently Bought Together