ContactUsLinkComponent
Amine-reactive Thermo Scientific TMT11-131C increases sample multiplexing from 10-plex to 11-plex, enabling even greater throughput for protein identification and quantitative analysis by tandem mass spectrometry (MS) 1 * 5 mg
Catalog # PIERA34807
Amine-reactive Thermo Scientific TMT11-131C increases sample multiplexing from 10-plex to 11-plex, enabling even greater throughput for protein identification and quantitative analysis by tandem mass spectrometry (MS) 1 * 5 mg
Catalog # PIERA34807
Frequently Bought Together