CARTRIDGE REPLACEMENT PR FLOWMETER ADM 1 * 1 items
Catalog # HEWLG6692A
Supplier: Agilent
Frequently Bought Together
VWR® Disposable Spatulas, PP
Supplier: VWR Collection
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Laboratory Markers
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium acetate trihydrate USP, Ph. Eur., BP
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Measuring cylinders, tall form, borosilicate glas, Class B, DURAN®
Supplier: DWK Life Sciences
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
1-Butanol, Supelco®
Supplier: Merck