Diethylene glycol dimethyl ether, 99%, stab. with 100ppm BHT 1 * 2 L
A13397.K3
Diethylene glycol dimethyl ether, 99%, stab. with 100ppm BHT 1 * 2 L
A13397.K3
Frequently Bought Together
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
: PanReac AppliChem
Isobutyronitrile 99%
: Thermo Fisher Scientific
n-Octane ≥98%
: Thermo Fisher Scientific
N,N-Dimethylformamide 99+%, extra pure
: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
: PanReac AppliChem
tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
: Thermo Fisher Scientific
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
Nitrosium tetrafluoroborate 97%
: Thermo Fisher Scientific
Acetone, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
Toluene 99.5% ACS for spectroscopy
: Thermo Fisher Scientific
Dibutylamine for HPLC
: Thermo Fisher Scientific