Iron aluminide, 97.5% (metals basis) 1 * 250 g
47233.30
Frequently Bought Together
Water, extra pure, Deionized
: Thermo Fisher Scientific
Acetonitrile, extra dry 99.9+%, AcroSeal®
: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.8% ACS for spectroscopy
: Thermo Fisher Scientific
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
2-Butoxyethanol ≥99%
: Thermo Fisher Scientific
Cerium(IV) oxide ≥99.5% (REO, rare earth oxide basis)
: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 124,000 99-100%, hydrolysed
: Thermo Fisher Scientific
1-Pentanol 99+% ACS
: Thermo Fisher Scientific
Toluene 99.8+% for analysis
: Thermo Fisher Scientific
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
: Honeywell Chemicals