ContactUsLinkComponent
VISOCOLOR Powder Pillows total Chlorine reagent set for photometric determination measuring range: 0.03-6.00 mg/L Cl2 1000 determina tions 1 * 1.000 items
Catalog # MANA936221.1
Supplier:  MACHEREY-NAGEL
undefined
VISOCOLOR Powder Pillows total Chlorine reagent set for photometric determination measuring range: 0.03-6.00 mg/L Cl2 1000 determina tions 1 * 1.000 items
Catalog # MANA936221.1
Supplier:  MACHEREY-NAGEL