ContactUsLinkComponent
VarioPrep HPLC column VP 150/40 NUCLEODUR 100-10 C18 ec *** with adjustable Endfitting *** length: 150 mm, ID: 40 mm pack of 1 1 * 1 items
Catalog # MANA762303.400
Supplier:  MACHEREY-NAGEL
VarioPrep HPLC column VP 150/40 NUCLEODUR 100-10 C18 ec *** with adjustable Endfitting *** length: 150 mm, ID: 40 mm pack of 1 1 * 1 items
Catalog # MANA762303.400
Supplier:  MACHEREY-NAGEL
Frequently Bought Together