ContactUsLinkComponent
VISOCOLOR Powder Pillows total Chlorine reagent set for photometric determination measuring range: 0.03-6.00 mg/L Cl2 100 determinat ions 1 * 100 items
Catalog # MANA936221
Supplier:  MACHEREY-NAGEL
undefined
VISOCOLOR Powder Pillows total Chlorine reagent set for photometric determination measuring range: 0.03-6.00 mg/L Cl2 100 determinat ions 1 * 100 items
Catalog # MANA936221
Supplier:  MACHEREY-NAGEL