ContactUsLinkComponent
VarioPrep HPLC column VP 15/50 NUCLEODUR C18 ec length: 15 mm, ID: 50 mm pack of 1 - compatible with VarioPrep HPLC column with ID 50 mm or larger - requires VP guard column holder 50 mm (short version, p/n 718255) 1 * 1 items
Catalog # MANA762311.500
Supplier:  MACHEREY-NAGEL
VarioPrep HPLC column VP 15/50 NUCLEODUR C18 ec length: 15 mm, ID: 50 mm pack of 1 - compatible with VarioPrep HPLC column with ID 50 mm or larger - requires VP guard column holder 50 mm (short version, p/n 718255) 1 * 1 items
Catalog # MANA762311.500
Supplier:  MACHEREY-NAGEL
Frequently Bought Together