ContactUsLinkComponent
VarioPrep HPLC guard column cartridge (short) VP 15/32 NUCLEODUR C18 Gravity-SB Length: 15 mm, ID: 32 mm pack of 1 - compatible wit h VarioPrep HPLC column with ID 32 or 40 mm - requires VP guard column holder 32 mm (REF 718253) 1 * 1 items
Catalog # MANA762355.320
Supplier:  MACHEREY-NAGEL
VarioPrep HPLC guard column cartridge (short) VP 15/32 NUCLEODUR C18 Gravity-SB Length: 15 mm, ID: 32 mm pack of 1 - compatible wit h VarioPrep HPLC column with ID 32 or 40 mm - requires VP guard column holder 32 mm (REF 718253) 1 * 1 items
Catalog # MANA762355.320
Supplier:  MACHEREY-NAGEL
Frequently Bought Together